Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354263

Sigma-Aldrich

Corning® Matrigel® Matrigel Matrix HiProtein GFR Mouse 10mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

shipped in

dry ice

storage temp.

−20°C

General description

Corning® Matrigel® matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Application

Corning® Matrigel® Matrigel Matrix HiProtein GFR Mouse can be used for:
  • Cell Growth and differentiation[1]
  • Metabolism/toxicology studies[2]
  • Invasion assays
  • In vitro and in vivo angiogenesis assays
  • In vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice

Legal Information

Corning is a registered trademark of Corning, Inc.
Matrigel is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Human iPS cell model of type 3 long QT syndrome recapitulates drug-based phenotype correction
Daniela M et al
Basic Research in Cardiology (2016)
Sonia Iranpour et al.
Journal of nanobiotechnology, 19(1), 314-314 (2021-10-14)
Improving anti-cancer drug delivery performance can be achieved through designing smart and targeted drug delivery systems (DDSs). For this aim, it is important to evaluate overexpressed biomarkers in the tumor microenvironment (TME) for optimizing DDSs. Herein, we designed a novel
Eman F Khaleel et al.
Clinical and experimental pharmacology & physiology, 47(6), 1092-1102 (2020-02-20)
This study investigated if EX-527 has an anti-tumour effect in SKOV-3 and OVCAR-3 ovarian cancer (OC) cell lines and if this effect involves the SIRT1/NF-κB axis. Cells were cultured in the presence or absence of EX-527, a selective SIRT-1 inhibitor.
Adrenergic Stress Protection of Human iPS Cell-Derived Cardiomyocytes by Fast Kv7. 1 Recycling
Ilaria P et al.
Frontiers in Physiology, 8 (2017)
Attalla Farag El-Kott et al.
Clinical and experimental pharmacology & physiology, 46(10), 944-954 (2019-07-06)
Autophagy promotes cell survival or induces apoptosis in cancer cells. While SIRT1 and AMPK induce autophagy in both normal and cancer cells, Akt and mTOR can inhibit it. Calycosin, a methoxyisoflavone, protects against several types of solid tumours including colorectal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service