Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354259

Sigma-Aldrich

Corning® Laminin/Entactin High Conc Mouse 10 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

biological source

mouse (Engelbreth-Holm-Swarm lathrytic mouse tumor)

assay

>90% (SDS-PAGE)

mol wt

900 kDa

packaging

pkg of 10.5 mg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Lamb2(16779)

General description

Laminin is a major component of basement membranes. It has numerous biological activities including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.

Quality

Quality:
Purity >/ 90% by SDS-PAGE
Tested for ability to initiate neurite outgrowth of NG-108 Mouse neuroblastoma cells
Found negative for the presence of bacteria, fungi and mycoplasma.

Physical form

High concentration laminin/ entactin complex frozen in Dulbecco′s PBS

Preparation Note

Handling:
For a firm gel, a concentration of at least 3.5 mg/mL is recommended.

Other Notes

Source
Engelbreth-Holm-Swarm (EHS) mouse tumor

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service