Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354249

Sigma-Aldrich

Corning® Collagen I Rat Tail High Protein 100 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

biological source

rat tail (tendon)

assay

>90% (SDS-PAGE)

form

liquid (in 0.02N Acetic acid with a concentration range of 8-11 mg/ml)

packaging

pkg of 100 mg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

General description

Corning® Collagen I HC, rat tail, 100 mg, is supplied as a liquid in 0.02 N acetic acid with a concentration range of 8-11mg/mL. The product has been tested for its ability to promote the attachment and spreading of HT 1080 human fibrosarcoma cells. 1/pack, formerly Cat. #40040. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.

Quality

  • Purity > 90% by SDS-PAGE
  • Tested for its ability to promote the attachment and spreading of HT 1080 Human Fibrosarcoma cells.
  • Found negative for the presence of bacteria, fungi and mycoplasma.
  • Forms a firm gel up to a concentration of 0.3 mg/mL. Further dilution may decrease the rigidity of the gel as will the time from manufacture.

Physical form

Collagen IHC, rat tail - 0.02 N in Acetic Acid

Other Notes

Biological Source:
Rat tail tendon

Concentration range:

8-11mg/mL

Handling:
Rat Tail Collagen HC is used as a three dimensional gel and will form a firm gel up to a concentration of 0.3 mg/mL. Gelling procedures included with product.

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service