DEN1513
Density Standard (15 °C)
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.1963 g/mL at 15 °C
About This Item
grade
certified reference material
agency
according to ASTM® D1480
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
density
1.1963 g/mL at 15 °C
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.
Application
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
294.8 °F - closed cup
flash_point_c
146.0 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service