Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DEN1502

Density Standard (15 °C)

UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.7306 g/mL at 15 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

according to ASTM® D1480

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

density

0.7306 g/mL at 15 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Density Standards can be found here.

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Application

Used as a certified density standard to confirm the accuracy of density measuring instrument and/or procedure of analytical testing method.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

pictograms

FlameHealth hazard

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-22.0 °F

flash_point_c

-30 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service