DEN10002
Density Standard (100 °C)
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.8103 g/mL at 100 °C
About This Item
Recommended Products
grade
certified reference material
agency
according to ASTM® D1480
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
density
0.8103 g/mL at 100 °C
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Manufactured and verified in an ISO 17025 and ISO 17034 accredited laboratory, as per UKAS
- Certified density value following ASTM D1480
- Provided in a tamper-evident packaging to assure safe delivery
- Marked holographically to ensure authenticity
- Compliant to ASTM and IP test method protocols
- Accompanied with a certificate of analysis (CoA) having information on expiry date and storage conditions for an effective audit
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service