Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D7044

Supelco

p-(N,N-Dichlorosulfamyl)benzoic acid

analytical standard, ≥70% (TLC)

Synonym(s):

Halazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl2NO4S
CAS Number:
Molecular Weight:
270.09
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥70% (TLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

OC(=O)c1ccc(cc1)S(=O)(=O)N(Cl)Cl

InChI

1S/C7H5Cl2NO4S/c8-10(9)15(13,14)6-3-1-5(2-4-6)7(11)12/h1-4H,(H,11,12)

Inchi Key

XPDVQPODLRGWPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Drews
Pflugers Archiv : European journal of physiology, 409(3), 251-257 (1987-07-01)
The effects of chloramine-T, a substance which partially and irreversibly inhibits inactivation of the Na current (Wang 1984), on the on and off charge movement (Qon and Qoff) and on the relative position of the Qon-E and F-E curves were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service