Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1383

Sigma-Aldrich

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate

≥98%

Synonym(s):

1,2-Cyclohexanedinitrilotetraacetic acid, Chel-CD, DCTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O8 · H2O
CAS Number:
Molecular Weight:
364.35
Beilstein/REAXYS Number:
3222748
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98% (T)
≥98%

mp

213-216 °C (lit.)

suitability

suitable for suitability for complexometry

SMILES string

[H]O[H].OC(=O)CN(CC(O)=O)[C@@H]1CCCC[C@H]1N(CC(O)=O)CC(O)=O

InChI

1S/C14H22N2O8.H2O/c17-11(18)5-15(6-12(19)20)9-3-1-2-4-10(9)16(7-13(21)22)8-14(23)24;/h9-10H,1-8H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24);1H2/t9-,10-;/m1./s1

InChI key

VASZYFIKPKYGNC-DHTOPLTISA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Chel is a trademark of Ciba-Geigy AG

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly M Brothers et al.
Cornea, 33(10), 1083-1087 (2014-08-05)
This study was designed to measure the impact of bacterial biofilms on diffusion of an ocular therapeutic through silicone hydrogel bandage lenses in vitro. An assay was designed to study the passage of a commonly used steroid, dexamethasone, through silicone
Charles C Kim et al.
Journal of bacteriology, 186(14), 4694-4704 (2004-07-03)
Survival and replication in the intracellular environment are critical components of the ability of Salmonella enterica serovar Typhimurium to establish systemic infection in the murine host. Intracellular survival is mediated by a number of genetic loci, including Salmonella pathogenicity island
Silke Lehmann et al.
PloS one, 15(6), e0234154-e0234154 (2020-06-05)
Phytohormones mediate most diverse processes in plants, ranging from organ development to immune responses. Receptor protein complexes perceive changes in intracellular phytohormone levels and trigger a signaling cascade to effectuate downstream responses. The in planta analysis of elements involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service