Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D007

Supelco

R(−)-Chloroethylnorapomorphine hydrochloride

analytical standard

Synonym(s):

NCA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18ClNO2 · HCl
CAS Number:
Molecular Weight:
352.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥85%

form

solid

storage condition

desiccated
protect from light

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

off-white

solubility

0.1 M HCl: soluble
H2O: soluble
aqueous base: unstable

format

neat

storage temp.

2-8°C

SMILES string

Cl.Oc1ccc2C[C@H]3N(CCCl)CCc4cccc(c34)-c2c1O

InChI

1S/C18H18ClNO2.ClH/c19-7-9-20-8-6-11-2-1-3-13-16(11)14(20)10-12-4-5-15(21)18(22)17(12)13;/h1-5,14,21-22H,6-10H2;1H/t14-;/m1./s1

InChI key

ASPGECRWCJCPPO-PFEQFJNWSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Irreversible dopamine receptor alkylating agent

Reconstitution

Dissolve in oxygen-free boiled water containing 0.1% sodium metabisulfite or other antioxidants. Solutions should be freshly prepared.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J Kilpatrick et al.
European journal of pharmacology, 107(1), 71-78 (1984-12-15)
N-(chloroethyl)-norapomorphine (NCA) irreversibly inhibits striatal dopamine function, the binding of dopamine agonist ligands and dopamine-stimulated adenylate cyclase. A selective interaction of [3H]NCA with dopamine receptor sites would be of use in the characterisation and isolation of brain dopamine receptors. In
J Lehmann et al.
European journal of pharmacology, 90(4), 393-400 (1983-06-17)
(-)-N-(2-Chloroethyl)-norapomorphine [-)-NCA) inhibited in a concentration-dependent manner the electrically evoked [3H]acetylcholine release in slices of cat caudate. The inhibition by (-)-NCA was reversible and antagonized by the benzamide neuroleptic S-sulpiride. Although (-)-NCA is an irreversible antagonist at some behaviorally relevant
S A Cohen et al.
Journal of medicinal chemistry, 26(10), 1348-1353 (1983-10-01)
The rates and mechanism of solvolysis of (-)-N-(2-chloroethyl)norapomorphine (NCA, 1c) in aqueous solution have been examined by reversed-phase liquid chromatography (HPLC) to follow the levels of starting material and products. The first-order rate constants for aziridinium ion formation at 25
The effects of (-)N-(chloroethyl)-norapomorphine on [3H]-apomorphine binding to striatal dopamine receptors.
M W Goosey et al.
Biochemical pharmacology, 31(22), 3693-3694 (1982-11-15)
Dopamine autoreceptors differ pharmacologically from postsynaptic dopamine receptors: effects of (-)-N-(2-chloroethyl)-norapomorphine.
J Lehmann et al.
European journal of pharmacology, 77(1), 85-86 (1982-01-08)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service