Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRMUPPLU1

Pour Point Standard

ISO Guide 34 Certified, pkg of 250 mL

Synonym(s):

Diesel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 250 mL

manufacturer/tradename

Paragon Scientific Ltd

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

134.6 °F

flash_point_c

57 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service