Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRMUCFGO

CFPP Standard

CFPP IP 309; ASTM D 6371 (-27.1°C)

Synonym(s):

Cold Filter Plugging Point Standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Cold Filter Plugging Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to ISO 17034 under UKAS accreditation

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

140.0 °F

flash_point_c

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service