Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRMPPGO

Pour Point Standard

ASTM D 97 (-29.2 °C)

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

according to ASTM® D97

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

bp

141-462 °C

application(s)

petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Pour Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

140.0 °F

flash_point_c

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service