CRM981
Dioxins/Furans - Clay Loam
certified reference material, pkg of 10 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of 10 g
manufacturer/tradename
RTC CRM981-010
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
matrix material
storage temp.
2-8°C
General description
Product CRM981-10G has been discontinued. Product SQC016-10G may be an acceptable replacement.
Analyte
Description
2,3,7,8-Tetrachlorodibenzo-p-dioxin solution
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anna K Wójtowicz et al.
Neurotoxicity research, 35(1), 183-195 (2018-08-19)
Di-(2-ethylhexyl) phthalate (DEHP) is used as a plasticizer in various plastic compounds, such as polyvinyl chloride (PVC), and products including baby toys, packaging films and sheets, medical tubing, and blood storage bags. Epidemiological data suggest that phthalates increase the risk
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service