CRM919
Aroclor 1221 - Sandy Loam
certified reference material, pkg of 50 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
grade
certified reference material
composition
Aroclor 1221, 22.0 mg/kg (assigned value)
packaging
pkg of 50 g
manufacturer/tradename
RTC CRM919-050
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
matrix
soil
pH
7.19
application(s)
environmental
format
matrix material
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service