Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM860

Supelco

Pesticides - Loamy Sand 2

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM860-050

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

pH

7.48

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    α-1,2,3,4,5,6-Hexachlorocyclohexane
    β-BHC
    δ-BHC
    α-Chlordane
    γ-Chlordane
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane
    4,4′-DDE
    3′-Deoxythymidine
    Dieldrin
    Endosulfan sulfate
    Endrin
    Endrin aldehyde
    Endrin ketone
    Heptachlor
    Hexachlorobenzene
    Methoxychlor
    See All (16)

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service