CRM821
OP Pesticide - Sandy Loam 1
certified reference material, pkg of 50 g
About This Item
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of 50 g
manufacturer/tradename
RTC CRM821-050
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
matrix
soil
pH
8.17
application(s)
environmental
format
matrix material
storage temp.
2-8°C
General description
Application
Other Notes
Analyte
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service