Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8079

Supelco

Quality Control Standard 21

certified reference material, 100 μg/mL each component in 5% HNO3/tr Tartaric acid/tr HF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

agency

EPA 200.7(Rev. 4.4)

packaging

bottle of 125 mL

concentration

100 μg/mL each component in 5% HNO3/tr Tartaric acid/tr HF

format

multi-component solution

Analyte

Description

    As 100 μg/mL
    Be 100 μg/mL
    Ca 100 μg/mL
    Cd 100 μg/mL
    Co 100 μg/mL
    Cr 100 μg/mL
    Cu 100 μg/mL
    Fe 100 μg/mL
    Li 100 μg/mL
    Mg 100 μg/mL
    Mn 100 μg/mL
    Mo 100 μg/mL
    Ni 100 μg/mL
    Pb 100 μg/mL
    Sb 100 μg/mL
    Se 100 μg/mL
    Sr 100 μg/mL
    Ti 100 μg/mL
    Tl 100 μg/mL
    V 100 μg/mL
    Zn 100 μg/mL
    See All (21)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service