CRM8074
Instrument Calibration Standard 1
certified reference material, 5000 μg/mL each component in 5% HNO3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
grade
certified reference material
Quality Level
packaging
bottle of 125 mL
concentration
5000 μg/mL each component in 5% HNO3
format
multi-component solution
storage temp.
room temp
General description
Our Instrument Calibration Standard 4 may be used separately or mixed together for preparation of the analytical curve. When mixed these solutions will yield a standard containing all the elements in the Target Analyte List (TAL). Instruments must be calibrated daily, every 24 hours or each time the instrument is set up. Calibration standards must be prepared fresh for each analysis and discarded after use.
Analyte
Description
Ca 5000 μg/mL
K 5000 μg/mL
Mg 5000 μg/mL
Na 5000 μg/mL
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
supp_hazards
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service