Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8071

Supelco

Spike Sample Standard 1

certified reference material, in 5% HNO3/tr Tartaric acid/tr HF (varied conc.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

bottle of 125 mL

concentration

in 5% HNO3/tr Tartaric acid/tr HF (varied conc.)

format

multi-component solution

storage temp.

room temp

Other Notes

In the spike sample analysis a spike containing the required elements, in their respective required amounts, is added to the sample prior to addition of any reagents, digestion, distillation, etc. Information is then provided on the effects of the sample matrix and the entrie methodology. The Sigma-Aldrich spike standard, CRM8071, provides all the analytes required for the ICP-AES and the AA spike. Add 1.0 mL of CRM8071 to aqueous samples and 2.0 mL of CRM8071 to solid samples prior to digestion.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Ag 5 μg/mL
    Al 200 μg/mL
    As 200 μg/mL
    Ba 200 μg/mL
    Be 5 μg/mL
    Cd 5 μg/mL
    Co 50 μg/mL
    Cr 20 μg/mL
    Cu 25 μg/mL
    Fe 100 μg/mL
    Mn 50 μg/mL
    Ni 50 μg/mL
    Pb 50 μg/mL
    Sb 50 μg/mL
    Se 200 μg/mL
    Tl 200 μg/mL
    V 50 μg/mL
    Zn 50 μg/mL
    See All (18)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service