Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8017

Supelco

Instrumentation Calibration Standard 1

for ICP-MS, certified reference material, 20 μg/mL each component in 5% HNO3/tr Tartaric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

bottle of 125 mL

concentration

20 μg/mL each component in 5% HNO3/tr Tartaric acid

format

multi-component solution

storage temp.

room temp

General description

Use for preparation every two weeks or as needed. Dilute to the concentration appropriate for the instrument with equal parts of Nitric Acid Blank and Water Blank.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Ag 20 μg/mL
    Al 20 μg/mL
    As 20 μg/mL
    Ba 20 μg/mL
    Be 20 μg/mL
    Cd 20 μg/mL
    Co 20 μg/mL
    Cr 20 μg/mL
    Cu 20 μg/mL
    Mn 20 μg/mL
    Mo 20 μg/mL
    Ni 20 μg/mL
    Pb 20 μg/mL
    Sb 20 μg/mL
    Se 20 μg/mL
    Th 20 μg/mL
    Tl 20 μg/mL
    U 20 μg/mL
    V 20 μg/mL
    Zn 20 μg/mL
    See All (20)

pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service