Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8015

Supelco

Tuning Solution 2

for ICP-MS, certified reference material, 10 μg/mL each component in nitric acid (2%)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product CRM8015 is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

packaging

bottle of 125 mL

concentration

10 μg/mL each component in nitric acid (2%)

format

multi-component solution

storage temp.

room temp

General description

We recommend this product for ICP-MS instrument tuning and mass calibration prior to analysis. A dilution of 100-fold to 1,000-fold, depending on the sensitivity of the instrument, is suggested. Dilute with equal parts of Nitric Acid Blank and Water Blank to yield a 1% nitirc acid matrix.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Ba 10 μg/mL
    Be 10 μg/mL
    Ce 10 μg/mL
    Co 10 μg/mL
    In 10 μg/mL
    Mg 10 μg/mL
    Pb 10 μg/mL
    Rh 10 μg/mL
    U 10 μg/mL
    See All (9)

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service