CRM8014
Tuning Solution 1
for ICP-MS, certified reference material, 10 μg/mL each component in 2% HNO3/5% HCl
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
bottle of 125 mL
concentration
10 μg/mL each component in 2% HNO3/5% HCl
format
multi-component solution
storage temp.
room temp
General description
Other Notes
Analyte
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service