Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8012

Supelco

Multi-element Solutions Set for ICP-MS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

description

for ICP-MS

packaging

bottle of 7 × 125 mL

storage temp.

room temp

General description

Our set of seven multi-element solutions is designed to contain virtually every element in the mass spectrum for concentration verification checks.

Kit Components Only

Product No.
Description

  • Multi-element solution 1, for ICP-MS, certified reference material, 10 μg/mL each component in 5% HNO3

  • Multi-element Solution 2, for ICP-MS, certified reference material, 10 μg/mL each component in 5% HNO3

  • Multi-element Solution 3, for ICP-MS, certified reference material, 10 μg/mL each component in 10% HCl/1% HNO3

  • Multi-element Solution 4, for ICP-MS, certified reference material, 10 μg/mL each component

  • Ultra Pure Nitric Acid, 2%

  • Ultra High Pure HCl, 2 %

  • Water, ASTM Type 1, 18 megohm


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service