Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM8004

Supelco

Multi-element Solution 2A

for ICP-MS, certified reference material, 10 μg/mL each component in 5% HNO3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

bottle of 125 mL

concentration

10 μg/mL each component in 5% HNO3

format

multi-component solution

storage temp.

room temp

Features and Benefits

Designed to contain virtually every element in the mass spectrum for concentration verification checks.
Mercury is supplied as a separate solution (CRM8082) due to incompatibility with other elements.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Ag 10 μg/mL
    Al 10 μg/mL
    As 10 μg/mL
    Ba 10 μg/mL
    Be 10 μg/mL
    Ca 10 μg/mL
    Cd 10 μg/mL
    Co 10 μg/mL
    Cr 10 μg/mL
    Cs 10 μg/mL
    Cu 10 μg/mL
    Fe 10 μg/mL
    Ga 10 μg/mL
    Hg 10 μg/mL
    K 10 μg/mL
    Li 10 μg/mL
    Mg 10 μg/mL
    Mn 10 μg/mL
    Na 10 μg/mL
    Ni 10 μg/mL
    Pb 10 μg/mL
    Rb 10 μg/mL
    Se 10 μg/mL
    Sr 10 μg/mL
    Tl 10 μg/mL
    U 10 μg/mL
    V 10 μg/mL
    Zn 10 μg/mL
    See All (28)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service