CRM373
TPH Banded - Clay Soil
certified reference material, pkg of 100 g
About This Item
Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of 100 g
manufacturer/tradename
RTC CRM373-100
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
matrix
soil
application(s)
environmental
format
matrix material
storage temp.
2-30°C
General description
Application
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Carc. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service