Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM371

Supelco

TPH - Loamy Soil

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

feature

Certified TPH Concentration - 2400 mg/Kg
Certified by FC-FID/MS

packaging

pkg of 100 g

manufacturer/tradename

RTC CRM371-100

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specificity

Specification:
  • C9 to C12 Aliphatics 105±29.5mg/Kg
  • C12 to C16 Aliphatics 369±70.8 mg/Kg
  • C16 to C21 Aliphatics 317±78.1 mg/Kg
  • C21 to C40 Aliphatics 44.7±17.9 mg/Kg
  • C10 to C12 Aromatics 34.3±7.02 mg/Kg
  • C12 to C16 Aromatics 152±35.9 mg/Kg
  • C16 to C21 Aromatics 205±56.3 mg/Kg
  • C21 to C36 Aromatics 31.8±12.2 mg/Kg

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service