Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM172

Supelco

PAHs - Sandy Loam Soil

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 100 g

manufacturer/tradename

RTC CRM172-100

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Acenaphthene
    Acenaphthylene
    Anthracene
    Benzo[a]pyrene
    Benzo[b]fluoranthene
    Benzo[ghi]perylene
    Benzo[k]fluoranthene
    Chrysene
    Fluoranthene
    Fluorene
    Naphthalene
    Phenanthrene
    Pyrene
    See All (13)

pictograms

Health hazardEnvironment

signalword

Danger

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service