Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLNSOIL1

Supelco

Clean Sandy Soil

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

matrix

soil

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Application

Product is intended for use as a clean control and as a base for fortification when no suitable CRM is available for use in method validation, method development or matrix recovery applications. COA covers most paramaters of interest.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2 Inhalation

target_organs

Lungs

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service