Key Documents
CFO12UKC
Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids (UKAS ISO 17025 certified)
Size 600, Nom. Constant 20, Range 4,000 to 20,000 cSt
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This product is issued with a UKAS 17025 certificate of calibration
Choose from one of the most recent versions:
Certificates of Analysis (COA)