Key Documents
CFO07NC
Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids
Size 300, Nom. Constant 0.25, Range 50 to 200 cSt, (non certified)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product CFO07UKC
Choose from one of the most recent versions:
Certificates of Analysis (COA)