Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CCW4695

Millipore

Indicator F

Indicator Reagent, suitable for microbiology, Custom product only

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

Quality Level

form

liquid

shelf life

12 mo. (Expiry date on the label)

feature

ready-to-use

composition

sodium chloride, 0.10 g/L
thymol blue, 0.02 g/L

packaging

box of 10 bottles

storage condition

(Tightly closed)

parameter

100 mL filling volume

bottle volume

125 mL

color

bluish

application(s)

food and beverages
microbiology

storage temp.

2-25°C

General description

Indicator F solution is formulated according to the customer specifications.

Use the medium according to the intended purpose, samples, and validated methods

Application

Indicator reagent for specific use according to customer specifications.

Components

  • 1 box with 10 bottles of 125 mL
  • Injectable cap.

Other Notes

Using syringe needles with a diameter greater than 0.8 mm is not recommended.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service