Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C9220

Millipore

Casein yeast magnesium broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

solid

shelf life

limited shelf life, expiry date on the label

final pH

7.0±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

nonselective for Escherichia coli
nonselective for coliforms

Application

Recommended for the growth of recombinant strains of E. coli.

Components

Components in g/L:
Casein enzymic hydrolysate, 10.00
Yeast extract, 5.00
Sodium chloride, 5.00
Magnesium sulfate, 0.98

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F R Blattner et al.
Science (New York, N.Y.), 196(4286), 161-169 (1977-04-08)
The Charon lambda bacteriophages have been developed as vectors for cloning. Their construction incorporates mutations that make them simple to use and also greatly increases their safety for the biological containment of cloned recombinant DNA. Three of the Charon vector

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service