C2725000
Copovidone - reference spectrum
European Pharmacopoeia (EP)
About This Item
Recommended Products
grade
pharmaceutical primary standard
application(s)
pharmaceutical (small molecule)
format
reference spectrum
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service