Key Documents
BSIPSL1NC
BS/IP/SL Viscometer for Transparent Liquids
Size 1 Nom. Constant 0.01, Range 3.5 to 10 cSt, (non certified)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product BSIPSL1UKC
Choose from one of the most recent versions:
Certificates of Analysis (COA)