Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP387

Ephedrine hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(1R,2S)-(−)-Ephedrine hydrochloride, (1R,2S)-α-[1-(Methylamino)ethyl]benzyl alcohol hydrochloride, (1R,2S)-(−)-2-Methylamino-1-phenyl-1-propanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · HCl
CAS Number:
Molecular Weight:
201.69
Beilstein/REAXYS Number:
3630391
MDL number:
UNSPSC Code:
41116107

grade

pharmaceutical primary standard

manufacturer/tradename

BP

mp

216-220 °C (lit.)

format

neat

Storage temp.

2-8°C

SMILES string

Cl.CN[C@@H](C)[C@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10-;/m0./s1

Inchi Key

BALXUFOVQVENIU-GNAZCLTHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service