Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP014

3-Amino-4-propoxybenzoic acid

British Pharmacopoeia (BP) Reference Standard, supplied as 2-(diethylamino)ethyl ester

Synonym(s):

3-Amino-4-propoxybenzoic acid 2-(diethylamino)ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$263.00

$263.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
50 MG
$263.00

About This Item

Empirical Formula (Hill Notation):
C16H26N2O3
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$263.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

proxymetacaine

form

solid

quality

supplied as 2-(diethylamino)ethyl ester

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[N+H](CCOC(=O)c1cc(c(cc1)OCCC)N)(CC)CC

InChI

1S/C16H26N2O3.ClH/c1-4-10-20-15-8-7-13(12-14(15)17)16(19)21-11-9-18(5-2)6-3;/h7-8,12H,4-6,9-11,17H2,1-3H3;1H

InChI key

BFUUJUGQJUTPAF-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

3-Amino-4-propoxybenzoic acid BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Proxymetacaine Eye Drops
  • Proxymetacaine Hydrochloride
  • Packaging

    Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class

    11 - Combustible Solids

    wgk_germany

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service