Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR724D

Glass-ceramic (thermal conductivity and diffusivity)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

agency

BCR®

form

particles
rod

manufacturer/tradename

JRC

diam. × H

26.9 mm × 22 mm

application(s)

pharmaceutical (small molecule)

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Certified for thermal conductivity and thermal diffusivity.

Analysis Note

For more information please see:
BCR724D

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service