BCR666
Asbestos fibres in lung tissue (low content of amosite + crocidolite)
BCR®, certified reference material
About This Item
grade
certified reference material
agency
BCR®
manufacturer/tradename
JRC
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
pharmaceutical (small molecule)
format
neat
Analysis Note
BCR666
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Carc. 1A - STOT RE 1
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service