Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR597

Sewage sludge (Cr)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product BCR597 is not currently sold in your country. Contact Technical Service

grade

certified reference material

agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental
pharmaceutical (small molecule)

format

matrix material

Analysis Note

For more information please see:
BCR-597_cert
BCR-597_report

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service