Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR519

Anhydrous butter fat (triglycerides)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24
Product BCR519 is not currently sold in your country. Contact Technical Service

grade

certified reference material

agency

BCR®

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

shipped in

wet ice

storage temp.

−20°C

Analysis Note

For more information please see:
BCR519

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Triglycerides C-24 - C 54, free Cholesterol

Matrix Group: Dairy products and eggs
Sales restrictions may apply

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Miriana Durante et al.
Food chemistry, 294, 112-122 (2019-05-28)
The feasibility of producing durum wheat pasta enriched with a lipophilic phytocomplex, extracted using supercritical carbon dioxide (SC-CO2), from ripe pumpkin, as free oil or as ready-to-mix oil/α-cyclodextrins (α-CDs) powder, was explored. Four types of pasta were prepared: (i) control

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service