Certified for the analytes listed below. See certificate for values and more details Vitamins: 5-Methyltetrahydrofolic acid (5-MTHF), Folate (total), Lutein, Thiamine (Vitamin B1), Total α-carotene, total-Lycopene, Total-β-carotene, trans-Lycopene, trans-α-Carotene, trans-β-Carotene, Vitamin B6 (total pyridoxine), Zeaxanthin
Matrix Group: Fruits and Vegetables
Legal Information
BCR is a registered trademark of European Commission
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
The tomato fruits during different stages of ripening have been extensively characterized for nutritionally important bioactives; however, changes in fatty acid composition are not available. Thus, in the present study, changes in fatty acid, along with carotenoid and α-tocopherol, were
A promising micropropagation protocol has been systematically established and demonstrated for the enhanced production of carotenoids, tocopherol and fatty acids in shoot tissues of
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.