Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR447

Rapeseed (oil, moisture, volatiles)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
85151701

grade

certified reference material

agency

BCR®

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The rapeseed reference material is to be analyzed according to the standard solvent procedure EEC, FOSFA or ISO. It can be used to calibrate NMR methods for the analysis of seeds of the same variety.
Mass fraction (g/100 g):
′As is′ Oil : 41.99 ± 0.15; Moisture and volatiles 7.42 ± 0.07; Dry basis Oil 45.36 ± 0.15

Packaging

About 150 g of rapeseed in a specially laminated bag containing an oxygen absorber and sealed under dry argon.

Analysis Note

For more information please see:
BCR447

Other Notes

Certified for the analytes listed below. See certificate for values and more details
"Moisture, “As is” Oil, “Dry Basis” Oil"

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service