Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR431

Brussels sprouts (vitamins)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701

grade

certified reference material

agency

BCR®

manufacturer/tradename

JRC

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

shipped in

wet ice

storage temp.

−20°C

Analysis Note

For more information please see:
BCR431

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Vitamins: Vitamin C (total ascorbate), Niacin (Vitamin B3)

Matrix Group: Fruits and Vegetables

Legal Information

BCR is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service