Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR301RM

Mullite (lattice spacing, other parameters)

Synonym(s):

Mullite

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

agency

BCR®

manufacturer/tradename

JRC

application(s)

industrial hygiene
pharmaceutical (small molecule)

format

matrix material

SMILES string

O=[Si]=O.O=[Al]O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

General description

Indicative value for relative reflection intensity.

Analysis Note

For more information please see:
BCR301RM

Legal Information

BCR is a registered trademark of European Commission

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service