Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR146R

Sewage sludge (industrial origin) (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

BCR®

application(s)

environmental
pharmaceutical (small molecule)

format

matrix material

Analysis Note

For more information please see:
BCR146R

Legal Information

BCR is a registered trademark of European Commission

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeng-Yei Hseu
Bioresource technology, 95(1), 53-59 (2004-06-23)
The production and application of compost potentially contaminate the environment with heavy metals. The heavy metal contents of composts must be exactly determined. Therefore, this study aims to understand the heavy metal contents in composts, using different digestion methods and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service