BCR105
Gas oil (0.363 % S)
BCR®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
grade
certified reference material
agency
BCR®
manufacturer/tradename
JRC
application(s)
pharmaceutical (small molecule)
format
neat
Analysis Note
For more information please see:
BCR105
BCR105
Legal Information
BCR is a registered trademark of European Commission
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service