Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR092

10-Azabenzo[a]pyrene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$765.00

$765.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$765.00

About This Item

Empirical Formula (Hill Notation):
C19H11N
CAS Number:
Molecular Weight:
253.30
Beilstein/REAXYS Number:
14606
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$765.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1cnc2c(c1)cc3ccc4cccc5ccc2c3c45

InChI

1S/C19H11N/c1-3-12-6-7-14-11-15-5-2-10-20-19(15)16-9-8-13(4-1)17(12)18(14)16/h1-11H

InChI key

BYFIUFNRSNKODH-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR092

Legal Information

BCR is a registered trademark of European Commission

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service