BCR090A
Titanium (impurities)
BCR®, certified reference material, disc
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
UNSPSC Code:
41116107
grade
certified reference material
agency
BCR®
form
disc
manufacturer/tradename
JRC
application(s)
pharmaceutical (small molecule)
format
matrix material
General description
Certified for the mass fractions of B, Co, Cr, Cu, Fe, Mn, Mo and Ni.
Analysis Note
For more information please see:
BCR090A
BCR090A
Legal Information
BCR is a registered trademark of European Commission
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service