Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B9273

Supelco

Butethamate citrate salt

analytical standard

Synonym(s):

2-Diethylaminoethyl 2-phenylbutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO2 · C6H8O7
CAS Number:
Molecular Weight:
455.50
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCC(C(=O)OCCN(CC)CC)c1ccccc1

InChI

1S/C16H25NO2.C6H8O7/c1-4-15(14-10-8-7-9-11-14)16(18)19-13-12-17(5-2)6-3;7-3(8)1-6(13,5(11)12)2-4(9)10/h7-11,15H,4-6,12-13H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

VVNURWKSPYZZCI-UHFFFAOYSA-N

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kailong Li et al.
Cancer discovery, 10(5), 724-745 (2020-03-20)
Mutations in protein-coding genes are well established as the basis for human cancer, yet how alterations within noncoding genome, a substantial fraction of which contain cis-regulatory elements (CRE), contribute to cancer pathophysiology remains elusive. Here, we developed an integrative approach

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service