Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4147

Supelco

Benzoylecgonine tetrahydrate

drug standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19NO4 · 4 H2O
CAS Number:
Molecular Weight:
361.39
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Product B4147 is not currently sold in your country. Contact Technical Service

grade

drug standard

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

10-30°C

SMILES string

O.O.O.O.CN1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3ccccc3)C(O)=O

InChI

1S/C16H19NO4.H2O/c1-17-11-7-8-12(17)14(15(18)19)13(9-11)21-16(20)10-5-3-2-4-6-10;/h2-6,11-14H,7-9H2,1H3,(H,18,19);1H2/t11-,12+,13-,14+;/m0./s1

InChI key

FFQVHSAKNBCHHT-BHARVXRSSA-N

Application

Benzoylecgonine tetrahydrate may be used as an analytical reference standard for the quantification of benzoylecgonine in biological samples[1][2] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Major metabolite of cocaine.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development and validation of a liquid chromatography?mass spectrometry assay for the determination of opiates and cocaine in meconium
Pichini.S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 794(2), 281-292 (2003)
Segmental analysis for cocaine and metabolites by HPLC in hair of suspected drug overdose cases
Clauwaert.MK, et al.
Forensic Science International, 110(3), 157-166 (2000)
Nahla A Abdelshafi et al.
Talanta, 165, 619-624 (2017-02-06)
This study focused on quantitative detection of cocaine on Euro banknotes in Germany. A sensitive direct competitive immunoassay was developed and optimized with a limit of detection (LOD) of 5.6ng/L. Exhaustive cocaine extraction by solvent was tested using different methanol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service